Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Chlorothioanisole 95.0+%, TCI America™
SDP

Catalog No. C09125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09125ML 5 mL
C091225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C09125ML Supplier TCI America Supplier No. C09125ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7205-91-6
Molecular Formula C7H7ClS
Molecular Weight (g/mol) 158.64
MDL Number MFCD00000921
InChI Key LLSMWLJPWFSMCP-UHFFFAOYSA-N
Synonym chloromethyl phenyl sulfide, chloromethylthiobenzene, alpha-chlorothioanisole, phenyl chloromethyl sulfide, chloromethyl phenyl sulphide, phenylthio methyl chloride, chloromethyl thio benzene, sulfide, chloromethyl phenyl, chloromethyl phenyl sulfane, benzene, chloromethyl thio
PubChem CID 81623
IUPAC Name [(chloromethyl)sulfanyl]benzene
SMILES ClCSC1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 mL
UN Number 3532
Formula Weight 158.64
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Chlorothioanisole
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.