Learn More
Bioworld alpha-Chymotrypsin, 250 mg
Catalog No. 50199010
Encompass_Preferred
Specification:Grade: USP gradeForm: White crystalline powderChymotrypsin acitvity (dry basis): 1000 U/mgTrypsin: 1%Residue after ignition: 2.5%Loss on drying: 5%Solubility (water, 10,000 U/ml): Pass
Catalog No. 50-199-010 Supplier Bioworld Supplier No. 215600361
Specification:Grade: USP gradeForm: White crystalline powderChymotrypsin acitvity (dry basis): 1000 U/mgTrypsin: 1%Residue after ignition: 2.5%Loss on drying: 5%Solubility (water, 10,000 U/ml): Pass