Learn More
BioVendor, LLC ALPHA CROSSLAP CTX-I ELISA (96 wells)
SDP
Supplier:  BioVendor, LLC AC04F1
Encompass
Sandwich ELISA, HRP-labelled antibodyThe ALPHA CrossLaps (CTX-I) ELISA is an enzyme-linked immunosorbent assay for the quantification of non-isomerized fragments of C-terminal telopeptides of Type-I collagen (α CTX-I) in human urine. The ALPHA CrossLaps (CTX-I) ELISA is intended for use as an indicator of degradation of non-isomerised bone collagen and may be used as an aid in the identification of skeletal metastases in patients with breast and prostate cancer. ApplicationsUrineSample Requirements45 µL (excluding dead volume) 25 μL pre-diluted (1+7)Calibration Range0-1600 ng/mLLimit of Detection0.80 ng/mL ALPHA CrossLaps
Catalog No. 50-196-3835