Learn More
Selleck Chemical LLC -cyano-4-hydroxycinnamic acid(-CHCA) 200mg 28166-41-8
SDP
Catalog No. 501948189
Encompass_Preferred
4-Chloro-?-cyanocinnamic acid (?-CHCA) is a classic monocarboxylate transporters (MCT) inhibitor. ?-cyano-4-hydroxycinnamate (CHC) has a 10-fold selectivity for MCT1 compared to other MCTs. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8189 Supplier Selleck Chemical LLC Supplier No. S8612200MG
4-Chloro-?-cyanocinnamic acid (?-CHCA) is a classic monocarboxylate transporters (MCT) inhibitor. ?-cyano-4-hydroxycinnamate (CHC) has a 10-fold selectivity for MCT1 compared to other MCTs. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.