Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Alpha-Cyclodextrin

Catalog No. 779008100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-900-8100G 1 kg
77-900-81KG 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 77-900-8100G Supplier Sigma Aldrich Supplier No. 779008100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Alpha-Cyclodextrin

Specifications

CAS 7585-39-9
Melting Point 290°C to 300°C
Molecular Formula C42H70O35
Linear Formula C42H70O35
MDL Number MFCD00078139
Quantity 1 kg
Synonym Cavamax(R) W6 Pharma; alpha-Cyclodextrin; alpha-Schardinger dextrin; Cyclohexaamylose; Cyclomaltohexaose
Molecular Weight (g/mol) 1134.98 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.