Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Cyclopentylmandelic acid, 98%

Catalog No. AC250970050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC250970050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC250970050 Supplier Thermo Scientific Chemicals Supplier No. 250970050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 427-49-6 | C13H16O3 | 220.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 427-49-6
Molecular Formula C13H16O3
Molecular Weight (g/mol) 220.27
MDL Number MFCD00019296
InChI Key WFLUEQCOAQCQLP-UHFFFAOYSA-N
Synonym alpha-cyclopentylmandelic acid, cyclopentylphenylglycolic acid, cyclopentyl hydroxy phenylacetic acid, cyclopentyl-hydroxy-phenyl-acetic acid, cyclopentyl mandelic acid, alpha-cyclopentyl-dl-mandelic acid, a-cyclo pentyl mandelic acid, alpha-cyclopentylmandelicacid, 2-rs-cyclopentyl-2-hydroxy-2-phenylacetic acid alpha-cyclopentylmandelic acid, cyclopentylmandelic acid
PubChem CID 98283
IUPAC Name 2-cyclopentyl-2-hydroxy-2-phenylacetic acid
SMILES C1CCC(C1)C(C2=CC=CC=C2)(C(=O)O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 144.0°C to 150.0°C
Color White to Beige
Boiling Point 110.0°C (0.5 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in bases
Formula Weight 220.27
Percent Purity 98%
Physical Form Powder
Chemical Name or Material α-Cyclopentylmandelic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 207-047-8
Recommended Storage Corrosives

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.