Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Cyclopentylphenylacetic Acid 98.0+%, TCI America™
SDP

Catalog No. C080810G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C080810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C080810G Supplier TCI America Supplier No. C080810G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3900-93-4
Molecular Formula C13H16O2
Molecular Weight (g/mol) 204.269
MDL Number MFCD00001379
InChI Key BCJIDGDYYYBNNB-UHFFFAOYSA-N
Synonym cyclopentyl phenyl acetic acid, cyclopentylphenylacetic acid, alpha-phenylcyclopentaneacetic acid, alpha-phenylcyclopentylacetic acid, acetic acid, cyclopentyphenyl, acetic acid, cyclopentylphenyl, benzeneacetic acid, .alpha.-cyclopentyl, alpha-cyclopentylphenylacetic acid, .alpha.-phenylcyclopentaneacetic acid, cyclopentaneacetic acid, .alpha.-phenyl
PubChem CID 98014
IUPAC Name 2-cyclopentyl-2-phenylacetic acid
SMILES C1CCC(C1)C(C2=CC=CC=C2)C(=O)O

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 10 g
Formula Weight 204.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material alpha-Cyclopentylphenylacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.