Learn More
TARGETMOL CHEMICALS INC ALPHA-CYPERONE 25MG
SDP
Catalog No. 502579711
Encompass_Preferred
Also available in 5 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. alpha-Cyperone ((alpha)-Cyperone) is a promising inhibitor of Hla production by S. aureus and protects lung cells from this bacterium. 2. alpha-Cyperone significantly inhibited PGE2 production by suppressing the LPS-induced expression of inducible COX-2 at both the mRNA and the protein levels downregulated the production and mRNA expression of the inflammatory cytokine IL-6. 3. alpha-Cyperone inhibited actin cytoskeleton polymerization induced by APEC-O78 through down regulating the expression of Nck-2 Cdc42 and Rac1. purity: 99%
Catalog No. 50-257-9711 Supplier TARGETMOL CHEMICALS INC Supplier No. T5S198125MG
Also available in 5 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. alpha-Cyperone ((alpha)-Cyperone) is a promising inhibitor of Hla production by S. aureus and protects lung cells from this bacterium. 2. alpha-Cyperone significantly inhibited PGE2 production by suppressing the LPS-induced expression of inducible COX-2 at both the mRNA and the protein levels downregulated the production and mRNA expression of the inflammatory cytokine IL-6. 3. alpha-Cyperone inhibited actin cytoskeleton polymerization induced by APEC-O78 through down regulating the expression of Nck-2 Cdc42 and Rac1. purity: 99%