Learn More
Medchemexpress LLC Alpha-D-galactose | 3646-73-9 | ≥98.0% | 1 MG
SDP
Catalog No. 5000232962
Encompass_Preferred
Alpha-D-galactose is the alpha anomer of D-galactose, a naturally occurring aldohexose used as a biochemical reagent in research. It is found in bulb tissues of Lilium davidii and is reported to have antioxidant, antitumor, immunomodulatory, and anti-HIV activities.
  • Alpha anomer of D-galactose (C6H12O6), molecular weight 180.16 g·mol-1.
  • High purity (≥98.0%) suitable for analytical and biological research.
  • Available in small quantities for research applications (1 mg, 5 mg, 10 mg).
  • Reported biological activities include antioxidant and immunomodulatory effects.
Catalog No. 50-002-32962 Supplier Medchemexpress LLC Supplier No. HY1213701MG
Alpha-D-galactose is the alpha anomer of D-galactose, a naturally occurring aldohexose used as a biochemical reagent in research. It is found in bulb tissues of Lilium davidii and is reported to have antioxidant, antitumor, immunomodulatory, and anti-HIV activities.
  • Alpha anomer of D-galactose (C6H12O6), molecular weight 180.16 g·mol-1.
  • High purity (≥98.0%) suitable for analytical and biological research.
  • Available in small quantities for research applications (1 mg, 5 mg, 10 mg).
  • Reported biological activities include antioxidant and immunomodulatory effects.