Learn More
Medchemexpress LLC Alpha-D-glucopyranose, 2,3,4,6-tetraacetate 1-(2,2,2-trichloroethanimidate) | 74808-10-9 | MFCD07369653 | 492.69 g/mol | C16H20Cl3NO10 | 10 MG
SDP
Catalog No. 5000299084
Encompass_Preferred
Alpha-D-glucopyranose, 2,3,4,6-tetraacetate 1-(2,2,2-trichloroethanimidate) is an acetyl-protected sugar derivative used as a glycosyl donor in carbohydrate synthesis and glycosylation reactions. It is intended for research use and is supplied in small milligram-scale quantities. Molecular formula C16H20Cl3NO10; molecular weight 492.69 g/mol.
  • Acetyl-protected glycosyl donor for selective glycosylation reactions.
  • Suitable for carbohydrate synthesis and method development.
  • Provided in small milligram quantities for laboratory research.
  • Reported CAS number 74808-10-9 for unambiguous identification.
Catalog No. 50-002-99084 Supplier Medchemexpress LLC Supplier No. HY4158910MG
Alpha-D-glucopyranose, 2,3,4,6-tetraacetate 1-(2,2,2-trichloroethanimidate) is an acetyl-protected sugar derivative used as a glycosyl donor in carbohydrate synthesis and glycosylation reactions. It is intended for research use and is supplied in small milligram-scale quantities. Molecular formula C16H20Cl3NO10; molecular weight 492.69 g/mol.
  • Acetyl-protected glycosyl donor for selective glycosylation reactions.
  • Suitable for carbohydrate synthesis and method development.
  • Provided in small milligram quantities for laboratory research.
  • Reported CAS number 74808-10-9 for unambiguous identification.