Learn More
Medchemexpress LLC Alpha-D-glucopyranose, hydrate (1:1) | 14431-43-7 | 98% | 2500 G
SDP
Catalog No. 5000342475
Encompass_Preferred
Alpha-D-glucose hydrate is a monosaccharide and the most common form of glucose. It cannot be broken down into simpler sugars and is crucial for energy metabolism, serving as the primary energy source for many cells. It also acts as a building block for larger carbohydrates like starch and glycogen. The "α" prefix indicates the orientation of the hydroxyl group on the first carbon atom, and it exists in solution as a hydrate, bound to water molecules.
  • Cannot be broken down into simpler sugars
  • Crucial for energy metabolism
  • Primary energy source for many cells
  • Building block for larger carbohydrates like starch and glycogen
  • Exists as a hydrate, bound to water molecules
  • For research use only
Catalog No. 50-003-42475 Supplier Medchemexpress LLC Supplier No. 2500GHY128417A
Alpha-D-glucose hydrate is a monosaccharide and the most common form of glucose. It cannot be broken down into simpler sugars and is crucial for energy metabolism, serving as the primary energy source for many cells. It also acts as a building block for larger carbohydrates like starch and glycogen. The "α" prefix indicates the orientation of the hydroxyl group on the first carbon atom, and it exists in solution as a hydrate, bound to water molecules.
  • Cannot be broken down into simpler sugars
  • Crucial for energy metabolism
  • Primary energy source for many cells
  • Building block for larger carbohydrates like starch and glycogen
  • Exists as a hydrate, bound to water molecules
  • For research use only