Learn More
Medchemexpress LLC Alpha-D-Glucopyranoside, 6-thio-alpha-D-glucopyranosyl 6-thio- | 1285607-08-0 | 99.9% | 374.43 | 25 MG
SDP
Catalog No. 5000391879
Encompass_Preferred
Fexlamose (AER-01) is a thiol-modified carbohydrate agent with mucolytic properties. It functions by cleaving disulfide bonds to cause mucolysis. This agent is used for research into chronic obstructive pulmonary disease (COPD) and muco-obstructive lung diseases (MOLD).
  • Thiol-modified carbohydrate agent.
  • Cleaves disulfides to cause mucolysis.
  • Applicable for research in chronic obstructive pulmonary disease (COPD).
  • Applicable for research in muco-obstructive lung diseases (MOLD).
  • In-vivo studies show it decreases airway mucus plugging.
  • In-vivo studies show it lessens airway inflammation.
  • In-vivo studies show it improves survival in muco-obstructive lung disease.
Catalog No. 50-003-91879 Supplier Medchemexpress LLC Supplier No. 25MGHY168973
Fexlamose (AER-01) is a thiol-modified carbohydrate agent with mucolytic properties. It functions by cleaving disulfide bonds to cause mucolysis. This agent is used for research into chronic obstructive pulmonary disease (COPD) and muco-obstructive lung diseases (MOLD).
  • Thiol-modified carbohydrate agent.
  • Cleaves disulfides to cause mucolysis.
  • Applicable for research in chronic obstructive pulmonary disease (COPD).
  • Applicable for research in muco-obstructive lung diseases (MOLD).
  • In-vivo studies show it decreases airway mucus plugging.
  • In-vivo studies show it lessens airway inflammation.
  • In-vivo studies show it improves survival in muco-obstructive lung disease.