Learn More
Medchemexpress LLC Alpha-D-glucose-13C | 287100-64-5 | 99.4% | 100 MG
SDP
Catalog No. 5000339768
Encompass_Preferred
Alpha-D-glucose-13C is a 13C labeled alpha-D-glucose, which is an endogenous metabolite. It is intended for research use only, not for medical applications.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes as tracers for quantitation during drug development
Catalog No. 50-003-39768 Supplier Medchemexpress LLC Supplier No. HY128417S1100MG
Alpha-D-glucose-13C is a 13C labeled alpha-D-glucose, which is an endogenous metabolite. It is intended for research use only, not for medical applications.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes as tracers for quantitation during drug development