Learn More
Medchemexpress LLC Alpha-D-glucose-13C | 287100-64-5 | 99.4% | 25 MG
SDP
Catalog No. 5000342975
Encompass_Preferred
Alpha-D-glucose-13C is a 13C labeled form of alpha-D-glucose, an endogenous metabolite. This compound is intended for research use only, serving as a valuable tool in scientific studies.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-42975 Supplier Medchemexpress LLC Supplier No. 25MGHY128417S1
Alpha-D-glucose-13C is a 13C labeled form of alpha-D-glucose, an endogenous metabolite. This compound is intended for research use only, serving as a valuable tool in scientific studies.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS