Learn More
Medchemexpress LLC Alpha-D-glucose-13C | 287100-64-5 | 99.4% | 50 MG
SDP
Catalog No. 5000338164
Encompass_Preferred
Alpha-D-glucose-13C is the 13C labeled alpha-D-glucose, an endogenous metabolite. Stable heavy isotopes, such as carbon-13, are incorporated into drug molecules for use as tracers in quantitation during drug development. This compound is for research use only.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules
  • Potential to affect pharmacokinetic and metabolic profiles of drugs (deuteration)
Catalog No. 50-003-38164 Supplier Medchemexpress LLC Supplier No. HY128417S150MG
Alpha-D-glucose-13C is the 13C labeled alpha-D-glucose, an endogenous metabolite. Stable heavy isotopes, such as carbon-13, are incorporated into drug molecules for use as tracers in quantitation during drug development. This compound is for research use only.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules
  • Potential to affect pharmacokinetic and metabolic profiles of drugs (deuteration)