Learn More
Medchemexpress LLC Alpha-D-glucose-13C | 287100-64-5 | MFCD08459778 | 99.4% | 181.15 g/mol | 13CC5H12O6 | 5 MG
SDP
Catalog No. 5000263423
Encompass_Preferred
Alpha-D-glucose-13C is a 13C-stable-isotope labeled form of alpha-D-glucose intended for research and analytical applications. It is used as a tracer and as an internal standard for quantitative studies in NMR, GC-MS, and LC-MS, and is supplied as a high-purity solid suitable for laboratory workflows.
  • 13C-labeled tracer useful for metabolic flux and pathway studies.
  • Suitable as an internal standard for quantitative NMR, GC-MS, and LC-MS assays.
  • High chemical purity (99.4%).
  • Available in small, precise package sizes for analytical workflows.
  • White to off-white solid, soluble in water for preparation of aqueous solutions.
Catalog No. 50-002-63423 Supplier Medchemexpress LLC Supplier No. HY128417S15MG
Alpha-D-glucose-13C is a 13C-stable-isotope labeled form of alpha-D-glucose intended for research and analytical applications. It is used as a tracer and as an internal standard for quantitative studies in NMR, GC-MS, and LC-MS, and is supplied as a high-purity solid suitable for laboratory workflows.
  • 13C-labeled tracer useful for metabolic flux and pathway studies.
  • Suitable as an internal standard for quantitative NMR, GC-MS, and LC-MS assays.
  • High chemical purity (99.4%).
  • Available in small, precise package sizes for analytical workflows.
  • White to off-white solid, soluble in water for preparation of aqueous solutions.