Learn More
Medchemexpress LLC Alpha-D-glucose hydrate | 14431-43-7 | 98% | 1 ML
SDP
Catalog No. 5000339120
Encompass_Preferred
alpha-D-glucose hydrate is a monosaccharide, the most common form of glucose, and cannot be broken down into simpler sugars. It plays a vital role in energy metabolism as a primary energy source for many cells and is a building block for larger carbohydrates like starch and glycogen. The α prefix indicates the orientation of the hydroxyl group on the first carbon atom, and it exists in solution as a hydrate.
  • Most common form of glucose
  • Plays a vital role in energy metabolism
  • Primary energy source for many cells
  • Building block of larger carbohydrates
  • Exists in solution as a hydrate
  • Biochemical reagent for life science related research
  • High purity of 98%
  • Soluble in DMSO at 100 mg/mL
Catalog No. 50-003-39120 Supplier Medchemexpress LLC Supplier No. HY128417A10MM/1ML
alpha-D-glucose hydrate is a monosaccharide, the most common form of glucose, and cannot be broken down into simpler sugars. It plays a vital role in energy metabolism as a primary energy source for many cells and is a building block for larger carbohydrates like starch and glycogen. The α prefix indicates the orientation of the hydroxyl group on the first carbon atom, and it exists in solution as a hydrate.
  • Most common form of glucose
  • Plays a vital role in energy metabolism
  • Primary energy source for many cells
  • Building block of larger carbohydrates
  • Exists in solution as a hydrate
  • Biochemical reagent for life science related research
  • High purity of 98%
  • Soluble in DMSO at 100 mg/mL