Learn More
Medchemexpress LLC Alpha-D-glucose pentaacetate | 604-68-2 | MFCD00064071 | >99.0% | 390.34 g/mol | C16H22O11 | 1 KG
SDP
Catalog No. 5000210621
Encompass_Preferred
α-D-Glucose pentaacetate (penta-O-acetyl-α-D-glucopyranose) is an acetylated derivative of D-glucose used as a protected sugar and intermediate in organic synthesis. It is commonly employed in glycosylation reactions and multi-step carbohydrate syntheses.
  • Acetylated protected sugar for organic synthesis.
  • Used as a glycosylation reagent and intermediate.
  • Solid reagent with molecular weight 390.34 g/mol.
  • CAS number 604-68-2 for unambiguous identification.
  • Commonly supplied in 1 kg packages.
Catalog No. 50-002-10621 Supplier Medchemexpress LLC Supplier No. HYW0497061KG
α-D-Glucose pentaacetate (penta-O-acetyl-α-D-glucopyranose) is an acetylated derivative of D-glucose used as a protected sugar and intermediate in organic synthesis. It is commonly employed in glycosylation reactions and multi-step carbohydrate syntheses.
  • Acetylated protected sugar for organic synthesis.
  • Used as a glycosylation reagent and intermediate.
  • Solid reagent with molecular weight 390.34 g/mol.
  • CAS number 604-68-2 for unambiguous identification.
  • Commonly supplied in 1 kg packages.