Learn More
Medchemexpress LLC Alpha-D-glucose (Standard) | 492-62-6 | 99.9% | 10 MG
SDP
Catalog No. 5000342044
Encompass_Preferred
Alpha-D-glucose (Standard) is the analytical standard of alpha-D-glucose. This monosaccharide has a more significant insulin stimulating effect than beta-D-glucose. It is intended for research and analytical applications, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS, serving as a reference standard.
  • Analytical standard for alpha-D-glucose
  • Monosaccharide
  • Used in qualitative, quantitative, and methodological research
  • Applied in HPLC, GC, and MS experiments
  • Intended for research and analytical applications
Catalog No. 50-003-42044 Supplier Medchemexpress LLC Supplier No. HY128417R10MG
Alpha-D-glucose (Standard) is the analytical standard of alpha-D-glucose. This monosaccharide has a more significant insulin stimulating effect than beta-D-glucose. It is intended for research and analytical applications, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS, serving as a reference standard.
  • Analytical standard for alpha-D-glucose
  • Monosaccharide
  • Used in qualitative, quantitative, and methodological research
  • Applied in HPLC, GC, and MS experiments
  • Intended for research and analytical applications