Learn More
Medchemexpress LLC Alpha-D-glucose (standard) | 492-62-6 | 99.9% | 25 MG
SDP
Catalog No. 5000343057
Encompass_Preferred
Alpha-D-glucose (Standard) is the analytical standard for alpha-D-glucose, intended for research and analytical applications. This monosaccharide has a more significant insulin stimulating effect than beta-D-glucose.
  • It is an analytical standard.
  • It is a reference standard supplied assay.
  • It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • It is classified as a saccharide and a monosaccharide.
  • Its initial source is microorganisms.
  • It is an endogenous metabolite.
  • It has a purity of 99.94%.
Catalog No. 50-003-43057 Supplier Medchemexpress LLC Supplier No. 25MGHY128417R
Alpha-D-glucose (Standard) is the analytical standard for alpha-D-glucose, intended for research and analytical applications. This monosaccharide has a more significant insulin stimulating effect than beta-D-glucose.
  • It is an analytical standard.
  • It is a reference standard supplied assay.
  • It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • It is classified as a saccharide and a monosaccharide.
  • Its initial source is microorganisms.
  • It is an endogenous metabolite.
  • It has a purity of 99.94%.