Learn More
Medchemexpress LLC Alpha-D-glucose (standard) | 492-62-6 | 99.9% | 50 MG
SDP
Catalog No. 5000339070
Encompass_Preferred
alpha-D-glucose (Standard) is the analytical standard of alpha-D-glucose. It is intended for research and analytical applications. Alpha-D-glucose is a monosaccharide that has a more significant insulin stimulating effect than beta-D-glucose.
  • Analytical standard used as a reference for assays
  • Commonly utilized in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Monosaccharide with a significant insulin stimulating effect
Catalog No. 50-003-39070 Supplier Medchemexpress LLC Supplier No. HY128417R50MG
alpha-D-glucose (Standard) is the analytical standard of alpha-D-glucose. It is intended for research and analytical applications. Alpha-D-glucose is a monosaccharide that has a more significant insulin stimulating effect than beta-D-glucose.
  • Analytical standard used as a reference for assays
  • Commonly utilized in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Monosaccharide with a significant insulin stimulating effect