Learn More
Medchemexpress LLC Alpha-D-mannose pentaacetate | 4163-65-9 | MFCD00069798 | 97.0% | 390.34 g/mol | C16H22O11 | 50 G
SDP
Catalog No. 5000301958
Encompass_Preferred
α-D-Mannose pentaacetate is an acetylated sugar derivative used as a protected mannose building block in organic synthesis and carbohydrate chemistry. Supplied as a solid for research use only, it is commonly used in glycosylation reactions, synthetic studies, and analytical applications requiring an acetyl-protected mannose.
  • Acetyl-protected mannose suitable for glycosylation and synthesis.
  • High purity (97.0%) for reproducible results.
  • Solid form, convenient handling and storage.
  • Molecular formula C16H22O11 and molecular weight 390.34 g/mol.
  • Available in laboratory-scale pack sizes for research workflows.
Catalog No. 50-003-01958 Supplier Medchemexpress LLC Supplier No. HYW03989850G
α-D-Mannose pentaacetate is an acetylated sugar derivative used as a protected mannose building block in organic synthesis and carbohydrate chemistry. Supplied as a solid for research use only, it is commonly used in glycosylation reactions, synthetic studies, and analytical applications requiring an acetyl-protected mannose.
  • Acetyl-protected mannose suitable for glycosylation and synthesis.
  • High purity (97.0%) for reproducible results.
  • Solid form, convenient handling and storage.
  • Molecular formula C16H22O11 and molecular weight 390.34 g/mol.
  • Available in laboratory-scale pack sizes for research workflows.