Learn More
CPC Scientific H-Ala-Phe-Thr-Cys-His-Cys-Arg-Arg-Ser-Cys-Tyr-Ser-Thr-Glu-Tyr-Ser-Tyr-Gly-Thr-Cys-Thr-Val-Met-Gly-Ile-Asn-His-Arg-Phe-Cys-Cys-Leu-OH (trifluoroacetate salt) 0.1MG
SDP
Supplier:  CPC Scientific DEFS008A
Encompass
This peptide contain 6 conserved disulfide-linked cysteines. In vitro, it has a prominent antimicrobial activities against bacteria, fungi, and certain enveloped viruses. In addition, human and rabbit defensins exert potent cytotoxicity in vitro against various mammalian tumor cells.SEQUENCE: H-Ala-Phe-Thr-Cys-His-Cys-Arg-Arg-Ser-Cys-Tyr-Ser-Thr-Glu-Tyr-Ser-Tyr-Gly-Thr-Cys-Thr-Val-Met-Gly-Ile-Asn-His-Arg-Phe-Cys-Cys-Leu-OH (trifluoroacetate salt)(Cys4 and Cys31/Cys6 and Cys20/Cys10 and Cys30 Disulfide Bridges)ONE-LETTER SEQUENCE: AFTCHCRRSCYSTEYSYGTCTVMGINHRFCCL (Cys4 and Cys31/Cys6 and Cys20/Cys10 and Cys30 Disulfide Bridges)MOLECULAR FORMULA: C156H228N46O46S7MOLECULAR WEIGHT: 3708.27STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [136661-76-2]SYNONYMS: -Defensin 6RESEARCH AREA: AntimicrobialCATEGORY: Defensins REFERENCES:P.A.Raj et al., Biochem. J., 347, 633 (
Catalog No. 50-210-9196