Learn More
Medchemexpress LLC Alpha-demethylnaproxen | 23981-47-7 | 99.8% | 216.24 | C13H12O3 | 1 ML
SDP
Catalog No. 5000208985
Encompass_Preferred
α-Demethylnaproxen is the major metabolite of the NSAID nabumetone and is provided as an analytical standard and research reagent. It is available as a prepared 10 mM solution in DMSO (1 mL) for analytical applications, and as solid quantities for synthesis or reference. Typical product data include CAS 23981-47-7, formula C13H12O3, molecular weight 216.24 g·mol⁻1, and high chemical purity. Store powder at -20°C (3 years) or 4°C (2 years); store solutions at -80°C (6 months) or -20°C (1 month). Intended for research use only.
  • For research use only.
  • Major metabolite of nabumetone.
  • Supplied as 10 mM solution in DMSO (1 mL).
  • High purity (~99.8%).
  • Stable under recommended storage conditions.
  • Suitable as an analytical standard for method development.
Catalog No. 50-002-08985 Supplier Medchemexpress LLC Supplier No. HYW08689610MM/1ML
α-Demethylnaproxen is the major metabolite of the NSAID nabumetone and is provided as an analytical standard and research reagent. It is available as a prepared 10 mM solution in DMSO (1 mL) for analytical applications, and as solid quantities for synthesis or reference. Typical product data include CAS 23981-47-7, formula C13H12O3, molecular weight 216.24 g·mol⁻1, and high chemical purity. Store powder at -20°C (3 years) or 4°C (2 years); store solutions at -80°C (6 months) or -20°C (1 month). Intended for research use only.
  • For research use only.
  • Major metabolite of nabumetone.
  • Supplied as 10 mM solution in DMSO (1 mL).
  • High purity (~99.8%).
  • Stable under recommended storage conditions.
  • Suitable as an analytical standard for method development.