Learn More
TARGETMOL CHEMICALS INC ALPHA-DIFLUOROMETHYL-DL- 5MG
SDP
Catalog No. 502580219
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. (alpha)-(difluoromethyl)-DL-Arginine (RMI 71897) is an enzyma-activated irreversible inhibitor of arginine decarboxylase for E. coli (Ki = 800 uM) Pseudomonas aeruginosa and Klebsiella pneumoniae. At 0.01 mM it has been shown to prevent osmotic stress-induced increases in arginine decarboxylase activity and putrescine synthesis in oat leaf cells. When combined with a variety of polyamine analogues (alpha)-(difluoromethyl)-DL-Arginine inhibited the growth of Trypanosoma Crui in mammalian host cells at a minimum concentration of 10 mM and prevented the growth of Trypanosoma Crui in T-cell receptor alpha-deficient mouse models. purity: 99%
Catalog No. 50-258-0219 Supplier TARGETMOL CHEMICALS INC Supplier No. T354495MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. (alpha)-(difluoromethyl)-DL-Arginine (RMI 71897) is an enzyma-activated irreversible inhibitor of arginine decarboxylase for E. coli (Ki = 800 uM) Pseudomonas aeruginosa and Klebsiella pneumoniae. At 0.01 mM it has been shown to prevent osmotic stress-induced increases in arginine decarboxylase activity and putrescine synthesis in oat leaf cells. When combined with a variety of polyamine analogues (alpha)-(difluoromethyl)-DL-Arginine inhibited the growth of Trypanosoma Crui in mammalian host cells at a minimum concentration of 10 mM and prevented the growth of Trypanosoma Crui in T-cell receptor alpha-deficient mouse models. purity: 99%