Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical α-DNA MeC(Bz) amidite | 1 g | CAS 152581-56-1 | InChIKey AGMUHEZFYUECLW-MRVYDGAMSA-N

Catalog No. 502364901
Encompass

α-DNA MeC(Bz) amidite is a Amidite reagent (Subcategory: α-DNA Series) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. SDS available for reference.

Specifications
- CAS: 152581-56-1
- MDL: No data
- InChIKey: AGMUHEZFYUECLW-MRVYDGAMSA-N
- Molecular Weight: 847.949761998
- Molecular Formula: C47H54N5O8P
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3S,5S)-5-(4-benzamido-5-methyl-2-oxopyrimidin-1(2H)-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: O=C1N=C(C(C)=CN1[C@@H]2C[C@@H]([C@H](O2)COC(C3=CC=C(C=C3)OC)(C4=CC=C(C=C4)OC)C5=CC=CC=C5)OP(N(C(C)C)C(C)C)OCCC#N)NC(C6=CC=CC=C6)=O

Catalog No. 50-236-4901 Supplier STA Pharmaceutical Supplier No. STAAM106001GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

α-DNA MeC(Bz) amidite is a Amidite reagent (Subcategory: α-DNA Series) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. SDS available for reference.

Specifications
- CAS: 152581-56-1
- MDL: No data
- InChIKey: AGMUHEZFYUECLW-MRVYDGAMSA-N
- Molecular Weight: 847.949761998
- Molecular Formula: C47H54N5O8P
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3S,5S)-5-(4-benzamido-5-methyl-2-oxopyrimidin-1(2H)-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: O=C1N=C(C(C)=CN1[C@@H]2C[C@@H]([C@H](O2)COC(C3=CC=C(C=C3)OC)(C4=CC=C(C=C4)OC)C5=CC=CC=C5)OP(N(C(C)C)C(C)C)OCCC#N)NC(C6=CC=CC=C6)=O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.