Learn More
Medchemexpress LLC Alpha-estradiol | 57-91-0 | MFCD00064144 | 99.9% | 272.39 | C18H24O2 | 10 MG
SDP
Catalog No. 5000298860
Encompass_Preferred
Alpha-estradiol (analytical standard) is a reference material intended for analytical testing and method development. It is a weak estrogen and a 5α-reductase inhibitor supplied as a high-purity solid suitable for calibration, qualitative analyses, and quantitative assays in research and laboratory workflows.
  • Analytical reference standard for research and method validation.
  • High stated purity for accurate calibration (99.93%).
  • Supplied as a solid for convenient handling and storage.
  • Available in small pack sizes appropriate for analytical use (10 mg).
  • Applicable to studies of estrogenic activity and 5α-reductase inhibition.
Catalog No. 50-002-98860 Supplier Medchemexpress LLC Supplier No. HYB0141AR10MG
Alpha-estradiol (analytical standard) is a reference material intended for analytical testing and method development. It is a weak estrogen and a 5α-reductase inhibitor supplied as a high-purity solid suitable for calibration, qualitative analyses, and quantitative assays in research and laboratory workflows.
  • Analytical reference standard for research and method validation.
  • High stated purity for accurate calibration (99.93%).
  • Supplied as a solid for convenient handling and storage.
  • Available in small pack sizes appropriate for analytical use (10 mg).
  • Applicable to studies of estrogenic activity and 5α-reductase inhibition.