Learn More
Medchemexpress LLC Alpha-Estradiol | 57-91-0 | MFCD00064144 | 99.9% | 500 MG
SDP
Catalog No. 5000450772
Encompass_Preferred
Alpha-Estradiol is a weak estrogen and a 5α-reductase inhibitor which is used as a topical medication in the treatment of androgenic alopecia.
  • Acts as a weak estrogen
  • Functions as a 5α-reductase inhibitor
  • Suitable for topical medication in the treatment of androgenic alopecia
  • Purity of 99.88%
  • Inhibits testosterone metabolism
  • Attenuates LPS-induced inflammatory markers
  • Decreases malondialdehyde (MDA) levels
Catalog No. 50-004-50772 Supplier Medchemexpress LLC Supplier No. HYB0141A500MG
Alpha-Estradiol is a weak estrogen and a 5α-reductase inhibitor which is used as a topical medication in the treatment of androgenic alopecia.
  • Acts as a weak estrogen
  • Functions as a 5α-reductase inhibitor
  • Suitable for topical medication in the treatment of androgenic alopecia
  • Purity of 99.88%
  • Inhibits testosterone metabolism
  • Attenuates LPS-induced inflammatory markers
  • Decreases malondialdehyde (MDA) levels