Learn More
Selleck Chemical LLC Alpha-Estradiol
SDP
Supplier:  Selleck Chemical LLC S59105MG
Encompass_Preferred
Alpha-Estradiol ( -Estradiol 17 alpha-Estradiol Alfatradiol Epiestradiol Epiestrol Alora 17 -E2) a natural non-feminizing stereoisomer a hormonally almost inactive isomer of physiological 17 beta-estradiol (17 -E2) is a weak inhibitor of estrogen
Catalog No. 50-000-03924