Learn More
Medchemexpress LLC Alpha-Estradiol (Standard) | 57-91-0 | 99.93% | 25 MG
SDP
Catalog No. 5000406818
Encompass_Preferred
Alpha-Estradiol (Standard) is the analytical standard of Alpha-Estradiol, intended for research and analytical applications. It is a weak estrogen and a 5α-reductase inhibitor used as a topical medication in the treatment of androgenic alopecia.
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
Catalog No. 50-004-06818 Supplier Medchemexpress LLC Supplier No. 25MGHYB0141AR
Alpha-Estradiol (Standard) is the analytical standard of Alpha-Estradiol, intended for research and analytical applications. It is a weak estrogen and a 5α-reductase inhibitor used as a topical medication in the treatment of androgenic alopecia.
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS