Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Estradiol, TCI America™
SDP

Catalog No. E09191G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E09191G Supplier TCI America Supplier No. E09191G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 57-91-0
Molecular Formula C18H24O2
Molecular Weight (g/mol) 272.388
MDL Number MFCD00064144
InChI Key VOXZDWNPVJITMN-SFFUCWETSA-N
Synonym alpha-estradiol, 17alpha-estradiol, alfatradiol, epiestradiol, 17-alpha-estradiol, 3,17-dihydroxyestratriene, 17a-estradiol, estradiol-17alpha, epiestrol, 17-epiestradiol
PubChem CID 68570
ChEBI CHEBI:17160
IUPAC Name (8R,9S,13S,14S,17R)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
SMILES CC12CCC3C(C1CCC2O)CCC4=C3C=CC(=C4)O

Specifications

Melting Point 224°C
Color White-Yellow
Quantity 1 g
Formula Weight 272.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha-Estradiol
RTECSNumber KG3750000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.