Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ alpha-Fetoprotein, 100ug

Catalog No. ICN770931 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
ICN No.: 770931; Quantity: 100microgram
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN770931 ICN No.: 770931; Quantity: 100microgram
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN770931 Supplier MP Biomedicals™ Supplier No. 08770931

missing translation for 'validMainframeMsg'

Specifications

For Use With (Application) Cell Signaling
Formulation Liquid in 0.1M Phosphate Buffer, 0.15M NaCl, pH 7.4
Name alpha Fetoprotein
pH Range 7.4
Purification Method SDS-PAGE
Quantity 100 μg
Source Human Cord Serum
Storage Requirements 0° to -20°C
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.