Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Furil Dioxime 97.0+%, TCI America™
SDP

Catalog No. F00791G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F00791G 1 g
F00795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F00791G Supplier TCI America Supplier No. F00791G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23789-34-6
Molecular Formula C10H8N2O4
Molecular Weight (g/mol) 220.18
MDL Number MFCD00075315,MFCD00075315
InChI Key BRKZOASOLMUAQJ-UHFFFAOYSA-N
PubChem CID 5359431
IUPAC Name N-[1,2-bis(furan-2-yl)-2-nitrosoethenyl]hydroxylamine
SMILES ONC(=C(N=O)C1=CC=CO1)C1=CC=CO1

Specifications

Melting Point 192°C
Quantity 1 g
Formula Weight 220.18
Percent Purity ≥97.0% (W)
Physical Form Crystalline Powder
Chemical Name or Material alpha-Furil Dioxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.