Learn More
AdipoGen alpha-Galactosylceramide [alpha-Gal-Cer; KRN7000]
SDP
Supplier:  AdipoGen AGCN20013M001
Encompass_Preferred
alpha-Galactosylceramide [alpha-Gal-Cer; KRN7000], AdipoGen Life Sciences. Chemical. CAS: 158021-47-7. Formula: C50H99NO9. MW: 858.3. Synthetic. Originally isolated from the marine sponge Angelas mauritianus. Immunostimulant. Potent stimulator of natural killer T (NKT) cells. Specific ligand of the lipid-binding MHC class I-like protein CD1d in human and mouse NKT cells. Protects against LPS-induced shock. Potent antitumor compound.
Catalog No. 50-464-318