Learn More
AdipoGen alpha-Galactosylceramide
SDP
Supplier:  AdipoGen AGCN20013C250
Encompass_Preferred
Chemical. CAS 158021-47-7. Formula C50H99NO9. MW 858.3. Synthetic. Originally isolated from the marine sponge Angelas mauritianus. Immunostimulant. Potent stimulator of natural killer T NKT cells. Specific ligand of the lipid-binding MHC class I-like protein CD1d in human and mouse NKT cells. Protects against LPS-induced shock. Potent antitumor compound.
Catalog No. 50-256-2696