Learn More
Abcam alpha-Galactosylceramide, NKT cell stimulator, 250UG
Catalog No. 502583285
Encompass_Preferred
MW 858.3 Da, Purity >96%. Potent NKT cell stimulator. Selective CD1d ligand. Stimulates lymphocyte proliferation and enhances antibody production. Shows antiangiogenic, antitumor and potent immunoregulatory effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3285 Supplier Abcam Supplier No. AB144262250UG
MW 858.3 Da, Purity >96%. Potent NKT cell stimulator. Selective CD1d ligand. Stimulates lymphocyte proliferation and enhances antibody production. Shows antiangiogenic, antitumor and potent immunoregulatory effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement