Learn More
AdipoGen alpha-Galactosylceramide Analog 8 (500 µg)
SDP
Supplier:  AdipoGen AGCR13622C500
Encompass_Preferred
Chemical. CAS: 922727-14-8. Formula: C50H97N3O8. MW: 868.3. Synthetic. Immunostimulant. Potent stimulator of natural killer T (NKT) cells. Specific ligand of the lipid-binding MHC class I-like protein CD1d in human and mouse NKT cells. Potent antitumor compound. Stimulates the production of smaller amounts of IFN-gamma than alpha-GalCer, but induces equal or greater IL-4 secretion in mouse models.
Catalog No. 50-205-3992