Learn More
Medchemexpress LLC Alpha-Gliadin (43-49) (Gliadorphin) | 107936-65-2 | MFCD00080188 | 99.8% | 875.97 | 50 MG
SDP
Catalog No. 5000422578
Encompass_Preferred
Alpha-Gliadin (43-49), also known as Gliadorphin, is a peptide that serves as the major protein component of gluten. This product can be utilized in studies involving the immunological response to alpha-gliadin.
  • Primary protein component of gluten
  • Inhibits the production of lymphokine activities by mononuclear leukocytes
  • Can be used in immunological response research to alpha-gliadin
Catalog No. 50-004-22578 Supplier Medchemexpress LLC Supplier No. HYP199950MG
Alpha-Gliadin (43-49), also known as Gliadorphin, is a peptide that serves as the major protein component of gluten. This product can be utilized in studies involving the immunological response to alpha-gliadin.
  • Primary protein component of gluten
  • Inhibits the production of lymphokine activities by mononuclear leukocytes
  • Can be used in immunological response research to alpha-gliadin