Learn More
Sigma Aldrich Fine Chemicals Biosciences alpha Glucosidase Activity
Catalog No. 502901908
Encompass
Glucosidase hydrolyzes carbohydrates by acting on terminal non-reducing a(1A†4)-linked D-glucose residues with the release of D-glucose. Defects in glucosidase have been implicated in Pompe disease and diabetes.
Catalog No. 50-290-1908 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK1231KT
Glucosidase hydrolyzes carbohydrates by acting on terminal non-reducing a(1A†4)-linked D-glucose residues with the release of D-glucose. Defects in glucosidase have been implicated in Pompe disease and diabetes.