Learn More
TARGETMOL CHEMICALS INC ALPHA-GLUCOSIDASE-IN-23 5MG
SDP
Catalog No. 502579275
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (alpha)-Glucosidase-IN-23 is an orally active (alpha)-Glucosidase inhibitor. (alpha)-Glucosidase-IN-23 can decrease blood glucose by (alpha)-glucosidase inhibition (IC50= 4.48 uM). purity: 99%
Catalog No. 50-257-9275 Supplier TARGETMOL CHEMICALS INC Supplier No. T677285MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (alpha)-Glucosidase-IN-23 is an orally active (alpha)-Glucosidase inhibitor. (alpha)-Glucosidase-IN-23 can decrease blood glucose by (alpha)-glucosidase inhibition (IC50= 4.48 uM). purity: 99%