Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alpha guanosine | 15398-66-0 | 96.0% | 283.24 g/mol | C10H13N5O5 | 10 MG
SDP

Catalog No. 5000285861
Encompass_Preferred

Alpha guanosine is a purine nucleoside analog used in biochemical and pharmacological research to study nucleoside metabolism and antimetabolite activity. The product is supplied as a purified powder with documented physicochemical properties, recommended storage conditions, and solubility information to support preparation of stable stock solutions for laboratory use.

  • Purity: 95.96%
  • Molecular formula: C10H13N5O5
  • Molecular weight: 283.24 g/mol
  • Soluble in DMSO at 12.5 mg/mL with ultrasonic warming to 60°C
  • Powder recommended storage: -20°C, stable for up to 3 years
  • In-solution stability: -80°C for 6 months, -20°C for 1 month
  • Available in small milligram pack sizes for research applications

Catalog No. 50-002-85861 Supplier Medchemexpress LLC Supplier No. HY15453610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alpha guanosine is a purine nucleoside analog used in biochemical and pharmacological research to study nucleoside metabolism and antimetabolite activity. The product is supplied as a purified powder with documented physicochemical properties, recommended storage conditions, and solubility information to support preparation of stable stock solutions for laboratory use.

  • Purity: 95.96%
  • Molecular formula: C10H13N5O5
  • Molecular weight: 283.24 g/mol
  • Soluble in DMSO at 12.5 mg/mL with ultrasonic warming to 60°C
  • Powder recommended storage: -20°C, stable for up to 3 years
  • In-solution stability: -80°C for 6 months, -20°C for 1 month
  • Available in small milligram pack sizes for research applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.