Learn More
Medchemexpress LLC Alpha-Guanosine | 15398-66-0 | C10H13N5O5 | 1 MG
SDP
Catalog No. 5000376445
Encompass_Preferred
Alpha-Guanosine is a purine nucleoside analog known for its broad antitumor activity, especially against indolent lymphoid malignancies. Its anticancer mechanisms include the inhibition of DNA synthesis and the induction of apoptosis.
  • Purine nucleoside analog
  • Broad antitumor activity
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis
Catalog No. 50-003-76445 Supplier Medchemexpress LLC Supplier No. HY1545361MG
Alpha-Guanosine is a purine nucleoside analog known for its broad antitumor activity, especially against indolent lymphoid malignancies. Its anticancer mechanisms include the inhibition of DNA synthesis and the induction of apoptosis.
  • Purine nucleoside analog
  • Broad antitumor activity
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis