Learn More
CPC Scientific H-Asp-Leu-Thr-Phe-His-Leu-Leu-Arg-Glu-Met-Leu-Glu-Met-Ala-Lys-Ala-Glu-Gln-Glu-Ala-Glu-Gln-Ala-Ala-Leu-Asn-Arg-Leu-Leu-Leu-Glu-Glu-Ala-NH2 1MG
SDP
Supplier:  CPC Scientific CRFS004B
Encompass
CRF antagonist, lyophilized solid that acts as a potent antagonist to CRF receptors to block the behavioral and physiological responses to a stressor when administered in the CNS.ONE-LETTER SEQUENCE: DLTFHLLREMLEMAKAEQEAEQAALNRLLLEEA-NH2MOLECULAR FORMULA: C166H274N46O53S2MOLECULAR WEIGHT:3826.41STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [99658-03-4], SYNONYMS: α-Helical CRF (9-41)RESEARCH AREA: HormonalREFERENCES: J. Rivier et al., Science, 224, 889 (1984)
Catalog No. 50-210-9166