Learn More
CPC Scientific H-Phe-His-Leu-Leu-Arg-Glu-Met-Leu-Glu-Met-Ala-Lys-Ala-Glu-Gln-Glu-Ala-Glu-Gln-Ala-Ala-Leu-Asn-Arg-Leu-Leu-Leu-Glu-Glu-Ala-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CRFS005B
Encompass
ONE-LETTER SEQUENCE: FHLLREMLEMAKAEQEAEQAALNRLLLEEA-NH2MOLECULAR FORMULA: C152H251N43O47S2MOLECULAR WEIGHT:3497.06STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [158535-55-8], SYNONYMS: α-Helical CRF (12-41)RESEARCH AREA: Hormonal
Catalog No. 50-210-9168