Learn More
ECM Bioscience αII-Spectrin (a.a. 1171-1176), cleavage-specific Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience SP5601
Encompass_Preferred
This antibody was affinity purified using αII-spectrin (a.a. 1171-1176) peptide (without carrier). The antibody detects a 136 kDa* cleaved fragment corresponding to αII-Spectrin on SDS-PAGE immunoblots of mouse diaphram treated with thapsigargin. The antibody only detects cleaved αII-Spectrin and does not detect full length αII-Spectrin. The immunogen was A synthetic peptide (coupled to KLH) corresponding to amino acid residues 1171 to 1176 in human αII-spectrin. This sequence has high homology with similar regions in rat and mouse αII-spectrin, but is not found in other spectrin family members. Accession #Q13813. Applications = WB, E; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Alpha-II spectrin, Fodrin alpha chain, Spectrin, non-erythroid alpha subunit, SPTAN1, NEAS, SPTA2
Catalog No. 50-172-0892