Learn More
Medchemexpress LLC Alpha-isomethyl ionone | 127-51-5 | MFCD00034582 | >98.0% | 206.32 g/mol | C14H22O | 25 G
SDP
Catalog No. 5000282785
Encompass_Preferred
Alpha-isomethyl ionone is a synthetic fragrance and flavor ingredient with a violet-like, floral-woody aroma. It is a low-molecular-weight ketone used in perfumery, flavor formulations, and analytical research. The material is typically supplied as a high-purity liquid for laboratory and formulation use (chemical formula C14H22O; molar mass 206.32 g/mol).
  • Violet-like floral and woody odor profile for perfumery and flavors.
  • High purity (>98.0%) suitable for analytical and research applications.
  • Low molecular weight and liquid form simplify handling and blending.
  • Compatible with common solvents and formulation matrices.
  • Supplied with standard chemical identifiers and documentation for regulatory use.
Catalog No. 50-002-82785 Supplier Medchemexpress LLC Supplier No. HYW12774025G
Alpha-isomethyl ionone is a synthetic fragrance and flavor ingredient with a violet-like, floral-woody aroma. It is a low-molecular-weight ketone used in perfumery, flavor formulations, and analytical research. The material is typically supplied as a high-purity liquid for laboratory and formulation use (chemical formula C14H22O; molar mass 206.32 g/mol).
  • Violet-like floral and woody odor profile for perfumery and flavors.
  • High purity (>98.0%) suitable for analytical and research applications.
  • Low molecular weight and liquid form simplify handling and blending.
  • Compatible with common solvents and formulation matrices.
  • Supplied with standard chemical identifiers and documentation for regulatory use.