Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich alpha-Keto-gamma-(methylthio)butyric acid sodium salt

Catalog No. K6000500MG
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K6000500MG 500 mg
50-290-8182 100 mg
K60001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. K6000500MG Supplier Sigma Aldrich Supplier No. K6000500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

alpha-Keto-gamma-(methylthio)butyric acid sodium salt

Specifications

CAS 51828-97-8
Molecular Formula C5H7NaO3S
Linear Formula C5H7O3SNa
MDL Number MFCD00010511
Quantity 500 mg
Synonym 4-Methylthio-2-oxobutanoic acid sodium salt; 4-Methylthio-2-oxobutyric acid sodium salt; KMBA
Molecular Weight (g/mol) 170.16
Percent Purity ≥97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.