Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Alpha-Ketoglutaric Acid

Catalog No. 75890500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-890-500G 500 g
75-890-100G 100 g
75-890-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 75-890-500G Supplier Sigma Aldrich Supplier No. 75890500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Alpha-Ketoglutaric Acid

Specifications

CAS 328-50-7
Melting Point 114°C to 118°C
Molecular Formula C5H6O5
Linear Formula HOOCCH2CH2COCOOH
MDL Number MFCD00004165
Quantity 500 g
Synonym alpha-Ketoglutarate; alpha-Ketoglutaric acid; 2-Oxoglutaric acid; 2-Oxopentanedioic acid
Molecular Weight (g/mol) 146.1 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.