Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich alpha-Ketoglutaric acid disodium salt dihydrate

Catalog No. 75892100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-892-100G 100 g
75-892-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 75-892-100G Supplier Sigma Aldrich Supplier No. 75892100G
May include imposed supplier surcharges.
 Restricted Item
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

alpha-Ketoglutaric acid disodium salt dihydrate (2-Oxoglutaric acid disodium salt) is the hydrated disodium salt of 2-oxoglutaric acid.α-Ketoglutaric acid disodium salt dihydrate (2-Oxoglutaric acid disodium salt) may be used as starting reagent for the synthesis of deuteriated 2-oxo[3,3-2H2]glutarate disodium salt.

Specifications

CAS 305-72-6
Molecular Formula C5H4Na2O5 -+ 2H2O
Linear Formula NaOOCCH2CH2COCOONa -+ 2H2O
MDL Number MFCD00150702
Quantity 100 g
Synonym 2-Oxoglutaric acid disodium salt; Sodium alpha-ketoglutarate dibasic
Molecular Weight (g/mol) 226.09
Percent Purity ≥98.0% (dried material, NT)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.