Learn More
Sigma Aldrich Fine Chemicals Biosciences alpha L Iduronidase human
Catalog No. 502907769
Encompass
L-Iduronidase (IDUA) is mapped to human chromosome 4p16.3. The mature IDUA protein is glycosylated and comprises triosephosphate isomerase (TIM) barrel domain -sandwich helix-loop-helix region and an immunoglobin-like domain. L-Iduronidase is classified under glycoside hydrolase (GH) family 39.
Catalog No. 50-290-7769 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. I603410UG
L-Iduronidase (IDUA) is mapped to human chromosome 4p16.3. The mature IDUA protein is glycosylated and comprises triosephosphate isomerase (TIM) barrel domain -sandwich helix-loop-helix region and an immunoglobin-like domain. L-Iduronidase is classified under glycoside hydrolase (GH) family 39.